Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057125.1 | NP_476466.1 | 10 | 2062 | ||
GenBank mRNA | D63673.1 | - | 10 | 2062 |
Gene Symbol: LOC100911599 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244507.1 | XP_006244569.1 | 638 |