Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057135.1 | NP_476476.1 | 13 | 1696 | ||
XM_006254192.2 | XP_006254254.1 | 13 | 2285 | |||
XM_006254193.2 | XP_006254255.1 | 13 | 2285 | |||
XM_006254195.2 | XP_006254257.1 | 13 | 2285 | |||
XM_006254196.2 | XP_006254258.1 | 13 | 2285 | |||
XM_006254197.2 | XP_006254259.1 | 13 | 2284 | |||
XM_006254198.2 | XP_006254260.1 | 13 | 2284 | |||
XM_008771850.1 | XP_008770072.1 | 13 | 2285 | |||
XM_008771851.1 | XP_008770073.1 | 13 | 1312 | |||
GenBank mRNA | D87240.1 | - | 13 | 1696 | ||
D87241.1 | - | 13 | 1696 | |||
D87242.1 | - | 13 | 1696 | |||
D87243.1 | - | 13 | 1696 | |||
D87244.1 | - | 13 | 1696 | |||
D87245.1 | - | 13 | 1696 | |||
D87246.1 | - | 13 | 1696 | |||
D87247.1 | - | 13 | 1696 | |||
EU034674.1 | - | 13 | 1291 |