Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057139.2 | NP_476480.2 | 3 | 940 | ||
XM_006250311.2 | XP_006250373.1 | 3 | 1316 | |||
GenBank mRNA | BC072529.1 | - | 3 | 940 | 7096906 | |
D14048.1 | - | 3 | 1419 |