Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baiap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057196.1 | NP_476544.1 | 10 | 1237 | ||
XM_006247869.2 | XP_006247931.1 | 10 | 1300 | |||
XM_006247870.2 | XP_006247932.1 | 10 | 1300 | |||
XM_006247871.2 | XP_006247933.1 | 10 | 1180 | |||
XM_006247872.2 | XP_006247934.1 | 10 | 1180 | |||
XM_008768464.1 | XP_008766686.1 | 10 | 1300 | |||
GenBank mRNA | AY037934.1 | - | 10 | 1237 | ||
BC074009.1 | - | 10 | 1271 | 7099884 |
Gene Symbol: LOC102555951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247941.2 | XP_006248003.1 | 3 | 444 | ||
XM_006247942.2 | XP_006248004.1 | 3 | 324 |