Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175578.3 | NP_783168.1 | 4 | 761 | ||
XM_006244593.2 | XP_006244655.1 | 3 | 685 | |||
GenBank mRNA | AF459022.1 | - | 3 | 483 | ||
AF459023.1 | - | 4 | 603 | |||
BC086974.1 | - | 4 | 761 | 7193921 |