Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caskin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080690.2 | NP_542421.2 | 18 | 3197 | ||
XM_006245895.2 | XP_006245957.1 | 18 | 4473 | |||
XM_006245896.2 | XP_006245958.1 | 16 | 4260 | |||
XM_006245897.2 | XP_006245959.1 | 16 | 4239 | |||
GenBank mRNA | AF451975.1 | - | 18 | 3220 |