Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: As3mt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080890.1 | NP_543166.1 | 10 | 981 | ||
GenBank mRNA | AF393243.1 | - | 10 | 981 |