Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080905.2 | NP_543181.2 | 2 | 877 | ||
XM_006255211.2 | XP_006255273.1 | 2 | 5478 | |||
XM_006255213.2 | XP_006255275.1 | 2 | 1217 | |||
XM_008772345.1 | XP_008770567.1 | 3 | 918 | |||
GenBank mRNA | AB067814.1 | - | 2 | 736 | ||
AF389476.1 | - | 1 | 697 | |||
AY681965.1 | - | 1 | 697 |