Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas1r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130818.1 | NP_570831.1 | 1 | 109 | ||
XM_008764334.1 | XP_008762556.1 | 2 | 905 | |||
GenBank mRNA | AF456324.1 | - | 1 | 109 | ||
AY032620.1 | - | 1 | 109 |
Set Membership: |
G-protein coupled receptor |