Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133321.2 | NP_579855.1 | 3 | 305 | ||
XM_006248128.2 | XP_006248190.1 | 4 | 394 | |||
XM_006248129.2 | XP_006248191.1 | 4 | 531 | |||
XM_006248130.2 | XP_006248192.1 | 5 | 693 | |||
XM_006248131.2 | XP_006248193.1 | 4 | 534 | |||
XM_006248132.2 | XP_006248194.1 | 4 | 485 | |||
XM_006248133.2 | XP_006248195.1 | 3 | 269 | |||
XM_006248135.2 | XP_006248197.1 | 3 | 726 | |||
XM_006248137.2 | XP_006248199.1 | 2 | 547 | |||
XM_006248138.2 | XP_006248200.1 | 2 | 186 | |||
XM_006248139.2 | XP_006248201.1 | 4 | 542 | |||
XM_006248141.2 | XP_006248203.1 | 2 | 1416 | |||
XM_008768550.1 | XP_008766772.1 | 4 | 502 | |||
XM_008768551.1 | XP_008766773.1 | 4 | 456 | |||
XM_008768552.1 | XP_008766774.1 | 4 | 413 | |||
XM_008768553.1 | XP_008766775.1 | 3 | 2669 | |||
GenBank mRNA | AY028455.1 | - | 3 | 306 |