Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133322.1 | NP_579856.1 | 7 | 1076 | ||
XM_006235721.2 | XP_006235783.1 | 7 | 1160 | |||
XM_006235725.2 | XP_006235787.1 | 7 | 1160 | |||
XM_006235726.2 | XP_006235788.1 | 7 | 1159 | |||
XM_006235727.2 | XP_006235789.1 | 7 | 1159 | |||
XM_006235728.2 | XP_006235790.1 | 7 | 1159 | |||
XM_006235729.2 | XP_006235791.1 | 7 | 1159 | |||
XM_008762457.1 | XP_008760679.1 | 7 | 1160 | |||
XM_008762458.1 | XP_008760680.1 | 7 | 1160 | |||
XM_008762459.1 | XP_008760681.1 | 7 | 1160 | |||
XM_008762460.1 | XP_008760682.1 | 7 | 1160 | |||
XM_008762461.1 | XP_008760683.1 | 7 | 1160 | |||
XM_008762462.1 | XP_008760684.1 | 7 | 1159 | |||
GenBank mRNA | AF087453.2 | - | 7 | 1076 |