Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A1cf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133400.1 | NP_596891.1 | 2, 3 | 189 | ||
XM_006231272.2 | XP_006231334.1 | 2, 3 | 360 | |||
GenBank mRNA | AF290984.1 | - | 2, 3 | 189 | ||
AF442133.1 | - | 2, 3 | 189 | |||
AF442134.1 | - | 2, 3 | 189 | |||
AY028945.1 | - | 1, 2 | 87 |