Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081972.1 | NP_001075441.1 | 2 | 172 | ||
NM_001081973.1 | NP_001075442.1 | 2 | 195 | |||
NM_001081974.1 | NP_001075443.1 | 2 | 181 | |||
XM_006240621.2 | XP_006240683.1 | 2 | 765 | |||
XM_006240622.2 | XP_006240684.1 | 2 | 780 | |||
XM_006240623.2 | XP_006240685.1 | 2 | 766 | |||
XM_006240625.2 | XP_006240687.1 | 2 | 661 | |||
XM_006240626.2 | XP_006240688.1 | 2 | 660 | |||
XM_006240627.2 | XP_006240689.1 | 2 | 661 | |||
XM_006240628.2 | XP_006240690.1 | 2 | 661 | |||
XM_006240629.2 | XP_006240691.1 | 2 | 660 | |||
XM_006240630.2 | XP_006240692.1 | 2 | 661 | |||
GenBank mRNA | M75146.1 | - | 2 | 172 | ||
M75147.1 | - | 2 | 195 | |||
M75148.1 | - | 2 | 181 |