Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyatl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134330.2 | NP_599157.2 | 6 | 736 | ||
GenBank mRNA | AB019693.1 | - | 802 | |||
BC078701.1 | - | 6 | 736 | 7130696 |