Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147206.2 | NP_671739.2 | 4, 5 | 416 | ||
GenBank mRNA | BC086985.1 | - | 4, 5 | 416 | 7125835 | |
U60085.2 | - | 4, 5 | 392 |