Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134359.1 | NP_599186.1 | 5 | 527 | ||
XM_006230206.1 | XP_006230268.1 | 6 | 523 | |||
GenBank mRNA | BC091574.1 | - | 5 | 527 | 7135215 |