Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134378.2 | NP_599205.1 | 7, 8 | 1147 | ||
XM_008763486.1 | XP_008761708.1 | 7, 8 | 1719 | |||
XM_008763487.1 | XP_008761709.1 | 7, 8 | 1873 | |||
XM_008763488.1 | XP_008761710.1 | 7, 8 | 1328 | |||
GenBank mRNA | AF230072.1 | - | 7, 8 | 1148 |