Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134386.1 | NP_599213.1 | 2 | 2807 | ||
XM_006257264.2 | XP_006257326.1 | 9 | 1792 | |||
XM_006257265.2 | XP_006257327.1 | 9 | 2615 | |||
XM_006257267.2 | XP_006257329.1 | 8 | 2484 | |||
XM_008773392.1 | XP_008771614.1 | 11 | 1999 | |||
XM_008773393.1 | XP_008771615.1 | 10 | 1923 | |||
XM_008773394.1 | XP_008771616.1 | 11 | 2003 | |||
XM_008773395.1 | XP_008771617.1 | 11 | 2043 | |||
XM_008773396.1 | XP_008771618.1 | 9 | 2577 | |||
XM_008773397.1 | XP_008771619.1 | 9 | 1851 | |||
XM_008773398.1 | XP_008771620.1 | 9 | 1850 | |||
GenBank mRNA | AB051807.1 | - | 2 | 2807 |