Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF063102.1 | - | 13 | 2460 | ||
AF081148.1 | - | 14 | 2906 | |||
AF081149.1 | - | 14 | 2906 | |||
AF081150.1 | - | 14 | 2906 | |||
AF081151.1 | - | 14 | 2906 | |||
AF081152.1 | - | 14 | 2906 | |||
AF081153.1 | - | 14 | 2906 |