Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syngap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113409.2 | NP_001106880.2 | 7 | 1483 | ||
NM_181092.4 | NP_851606.3 | 9 | 1546 | |||
GenBank mRNA | AF048976.1 | - | 9 | 1554 | ||
AF050183.2 | - | 5 | 1235 | |||
AF058789.2 | - | 9 | 1433 | |||
AF058790.1 | - | 7 | 1301 |