Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc13d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138844.1 | NP_620199.1 | 11 | 1077 | ||
XM_006247702.2 | XP_006247764.1 | 11 | 1262 | |||
GenBank mRNA | AF159356.1 | - | 11 | 1077 |