Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Podxl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138848.1 | NP_620203.1 | 2 | 336 | ||
XM_008762758.1 | XP_008760980.1 | 2 | 496 | |||
GenBank mRNA | AB020726.1 | - | 2 | 409 | ||
AF109393.1 | - | 2 | 336 | |||
BC070886.1 | - | 2 | 406 | 7098164 |