Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138850.1 | NP_620205.1 | 9 | 830 | ||
GenBank mRNA | AF493782.1 | - | 9 | 830 | ||
BC061713.1 | - | 9 | 828 |