Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138853.2 | NP_620208.1 | 10 | 847 | ||
XM_006239000.1 | XP_006239062.1 | 10 | 909 | |||
XM_006239002.1 | XP_006239064.1 | 8 | 783 | |||
XM_006239004.1 | XP_006239066.1 | 8 | 763 | |||
XM_006239005.2 | XP_006239067.1 | 7 | 1117 | |||
XM_008764131.1 | XP_008762353.1 | 8 | 915 | |||
XM_008764132.1 | XP_008762354.1 | 8 | 956 | |||
GenBank mRNA | AB072920.1 | - | 9 | 775 | ||
BC078730.1 | - | 10 | 847 | 7134156 |