Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138897.1 | NP_620252.1 | 6 | 917 | ||
GenBank mRNA | D50671.1 | - | 6 | 917 |