Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138979.2 | NP_620435.1 | 4 | 763 | ||
GenBank mRNA | AJ421029.1 | - | 4 | 794 | ||
BC081725.1 | - | 4 | 763 | 7127281 |