Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Comt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012531.2 | NP_036663.1 | 5 | 796 | ||
XM_006248603.2 | XP_006248665.1 | 5 | 797 | |||
GenBank mRNA | BC081850.1 | - | 5 | 654 | 7191956 | |
FQ209401.1 | - | 4 | 509 | |||
FQ209488.1 | - | 4 | 515 | |||
FQ209638.1 | - | 4 | 509 | |||
FQ209706.1 | - | 4 | 520 | |||
FQ210267.1 | - | 4 | 509 | |||
FQ210416.1 | - | 4 | 537 | |||
FQ210430.1 | - | 4 | 512 | |||
FQ218190.1 | - | 4 | 511 | |||
FQ218375.1 | - | 4 | 519 | |||
FQ218564.1 | - | 4 | 511 | |||
FQ218595.1 | - | 4 | 511 | |||
FQ219155.1 | - | 4 | 516 | |||
FQ219257.1 | - | 4 | 516 | |||
FQ219433.1 | - | 4 | 511 | |||
FQ219709.1 | - | 2 | 162 | |||
FQ223536.1 | - | 4 | 511 | |||
FQ228829.1 | - | 4 | 509 | |||
FQ228870.1 | - | 4 | 517 | |||
FQ229523.1 | - | 4 | 491 | |||
M60753.1 | - | 600 |