Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270852.1 | NP_001257781.1 | 10 | 997 | ||
NM_001270853.1 | NP_001257782.1 | 9 | 891 | |||
NM_012545.4 | NP_036677.1 | 10 | 1037 | |||
XM_006251474.2 | XP_006251536.1 | 10 | 1773 | |||
XM_006251475.2 | XP_006251537.1 | 10 | 1613 | |||
XM_008770251.1 | XP_008768473.1 | 10 | 1068 | |||
XM_008770252.1 | XP_008768474.1 | 10 | 1131 | |||
XM_008770253.1 | XP_008768475.1 | 9 | 948 | |||
XM_008770254.1 | XP_008768476.1 | 10 | 1068 | |||
GenBank mRNA | BC087032.1 | - | 10 | 997 | 7098083 | |
FQ219697.1 | - | 10 | 1035 | |||
JF273828.1 | - | 9 | 891 | |||
M27716.1 | - | 10 | 1042 | |||
U31884.1 | - | 10 | 1019 |