Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750657.3 | XP_003750705.1 | 11 | 1327 | ||
XM_006244636.2 | XP_006244698.1 | 11 | 1559 |
Gene Symbol: Eno2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139325.3 | NP_647541.1 | 11 | 1271 | ||
XM_006237330.2 | XP_006237392.1 | 11 | 1557 | |||
GenBank mRNA | AF019973.1 | - | 11 | 1271 | ||
BC060310.1 | - | 11 | 1262 | 6888358 | ||
FQ214264.1 | - | 11 | 1246 | |||
M11931.1 | - | 11 | 1271 |