Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hprt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012583.2 | NP_036715.1 | 8 | 625 | ||
GenBank mRNA | BC098629.1 | - | 8 | 625 | 7386548 | |
FQ212770.1 | - | 209 | ||||
FQ212939.1 | - | 209 | ||||
FQ213155.1 | - | 209 | ||||
FQ215291.1 | - | 209 | ||||
FQ215779.1 | - | 209 | ||||
FQ216318.1 | - | 209 | ||||
FQ219798.1 | - | 209 | ||||
FQ219974.1 | - | 209 | ||||
FQ220004.1 | - | 209 | ||||
FQ220248.1 | - | 209 | ||||
FQ229708.1 | - | 209 | ||||
FQ230307.1 | - | 209 | ||||
FQ234297.1 | - | 209 | ||||
M63983.1 | - | 8 | 657 | |||
M86443.1 | - | 385 | ||||
S79292.1 | - | 8 | 657 | |||
X62085.1 | - | 585 |
Gene Symbol: LOC103689983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773659.1 | XP_008771881.1 | 8 | 701 |