Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nupl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139091.2 | NP_620791.1 | 7, 8 | 821 | ||
XM_006252093.2 | XP_006252155.1 | 7, 8 | 926 | |||
GenBank mRNA | AF000901.1 | - | 6, 7 | 714 | ||
BC085690.1 | - | 7, 8 | 821 | 7190650 | ||
U63839.1 | - | 7, 8 | 834 |