Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270788.1 | NP_001257717.1 | 7 | 931 | ||
NM_139100.2 | NP_620800.2 | 7 | 928 | |||
GenBank mRNA | BC070918.1 | - | 7 | 870 | 7104964 | |
FQ210190.1 | - | 7 | 882 | |||
FQ213208.1 | - | 2 | 94 | |||
FQ213570.1 | - | 2 | 94 | |||
FQ213750.1 | - | 7 | 887 | |||
FQ214401.1 | - | 2 | 94 | |||
FQ214581.1 | - | 2 | 94 | |||
FQ214698.1 | - | 7 | 885 | |||
FQ215697.1 | - | 7 | 843 | |||
FQ215957.1 | - | 7 | 887 | |||
FQ216084.1 | - | 2 | 94 | |||
FQ216349.1 | - | 7 | 884 | |||
FQ216361.1 | - | 7 | 885 | |||
FQ216409.1 | - | 7 | 892 | |||
FQ216421.1 | - | 7 | 885 | |||
FQ216474.1 | - | 7 | 893 | |||
FQ216716.1 | - | 2 | 94 | |||
FQ216978.1 | - | 2 | 94 | |||
FQ217343.1 | - | 2 | 94 | |||
FQ217707.1 | - | 7 | 885 | |||
FQ217883.1 | - | 2 | 112 | |||
FQ220537.1 | - | 7 | 882 | |||
FQ220626.1 | - | 7 | 869 | |||
FQ222611.1 | - | 2 | 94 | |||
FQ223916.1 | - | 7 | 885 | |||
FQ224211.1 | - | 2 | 94 | |||
FQ227446.1 | - | 7 | 882 | |||
FQ229113.1 | - | 7 | 882 | |||
FQ230627.1 | - | 7 | 884 | |||
FQ232293.1 | - | 5 | 687 | |||
FQ232835.1 | - | 7 | 878 | |||
FQ234854.1 | - | 7 | 882 | |||
M23984.1 | - | 6 | 847 |