Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012610.2 | NP_036742.2 | 6 | 1348 | ||
GenBank mRNA | X05137.1 | - | 6 | 1348 |