Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113747.1 | NP_001107219.1 | 812 | |||
XM_006248028.2 | XP_006248090.1 | 5110 | ||||
XM_008768559.1 | XP_008766781.1 | 7328 | ||||
XM_008768560.1 | XP_008766782.1 | 4106 | ||||
XM_008768561.1 | XP_008766783.1 | 3839 | ||||
GenBank mRNA | AF388528.1 | - | 1 | 185 | ||
BC079469.1 | - | 648 | ||||
BC158669.1 | - | 812 | 8370827 |