Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_341949.4 | XP_341950.2 | 1, 2 | 1354 | ||
GenBank mRNA | L13205.1 | - | 1, 2 | 204 |