Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100823.1 | NP_001094293.1 | 9 | 1564 | ||
XM_006257442.2 | XP_006257504.1 | 9 | 1751 | |||
GenBank mRNA | AF130121.1 | - | 9 | 1327 |