Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144561.2 | NP_653147.1 | 8 | 1283 | ||
NM_144562.1 | NP_653148.1 | 6 | 888 | |||
XM_006250144.2 | XP_006250206.1 | 8 | 1218 | |||
GenBank mRNA | AF458416.1 | - | 8 | 1284 | ||
AF458417.1 | - | 6 | 888 | |||
BC081721.1 | - | 8 | 1273 | 7107835 |