Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017040.1 | NP_058736.1 | 1 | 372 | ||
GenBank mRNA | BC085926.1 | - | 1 | 356 | 7109733 | |
M23591.1 | - | 1 | 372 | |||
X14087.1 | - | 1 | 205 | |||
X16044.1 | - | 1 | 107 |