Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145674.1 | NP_663707.1 | 7 | 1168 | ||
XM_006234033.2 | XP_006234095.1 | 7 | 1303 | |||
XM_006234037.2 | XP_006234099.1 | 7 | 1618 | |||
XM_008761700.1 | XP_008759922.1 | 7 | 1303 | |||
XM_008761701.1 | XP_008759923.1 | 6 | 1127 | |||
XM_008761702.1 | XP_008759924.1 | 5 | 950 | |||
GenBank mRNA | AF488784.1 | - | 6 | 925 | ||
AF520589.1 | - | 7 | 1168 | |||
BC166599.1 | - | 7 | 1303 | 7932138 | ||
FQ222249.1 | - | 7 | 1305 |