Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hint3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100825.1 | NP_001094295.1 | 2 | 227 | ||
NM_001276433.1 | NP_001263362.1 | 2 | 227 | |||
GenBank mRNA | AY040767.1 | - | 2 | 170 | ||
CB741212.1 | - | 2 | 84 | |||
FQ215151.1 | - | 2 | 227 |