Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109887.1 | NP_001103357.1 | 21 | 2140 | ||
NM_001109888.1 | NP_001103358.1 | 21 | 2119 | |||
NM_001109889.1 | NP_001103359.1 | 21 | 2116 | |||
NM_001109890.1 | NP_001103360.1 | 20 | 1993 | |||
NM_138507.2 | NP_612516.2 | 23 | 2389 | |||
XM_006249910.2 | XP_006249972.1 | 23 | 2483 | |||
XM_006249912.2 | XP_006249974.1 | 22 | 2317 | |||
XM_008769534.1 | XP_008767756.1 | 22 | 2320 | |||
GenBank mRNA | FQ230923.1 | - | 10 | 726 | ||
FQ232030.1 | - | 4 | 291 | |||
FQ232152.1 | - | 4 | 291 | |||
M10072.1 | - | 15 | 1732 | |||
Y00065.1 | - | 21 | 2218 |