Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109887.1 | NP_001103357.1 | 20 | 1968 | ||
NM_001109888.1 | NP_001103358.1 | 20 | 1947 | |||
NM_001109889.1 | NP_001103359.1 | 20 | 1944 | |||
NM_001109890.1 | NP_001103360.1 | 19 | 1821 | |||
NM_138507.2 | NP_612516.2 | 22 | 2217 | |||
XM_006249910.2 | XP_006249972.1 | 22 | 2311 | |||
XM_006249912.2 | XP_006249974.1 | 21 | 2145 | |||
XM_008769534.1 | XP_008767756.1 | 21 | 2148 | |||
GenBank mRNA | FQ230923.1 | - | 9 | 554 | ||
FQ232030.1 | - | 3 | 119 | |||
FQ232152.1 | - | 3 | 119 | |||
M10072.1 | - | 14 | 1560 | |||
Y00065.1 | - | 20 | 2046 |