Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012641.1 | NP_036773.1 | 4 | 326 | ||
GenBank mRNA | FQ232485.1 | - | 4 | 318 | ||
M18962.1 | - | 3 | 302 | |||
M62930.1 | - | 4 | 326 |