Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sparc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012656.1 | NP_036788.1 | 11 | 943 | ||
XM_008767663.1 | XP_008765885.1 | 10 | 1021 | |||
GenBank mRNA | BC061777.1 | - | 10 | 958 | 5621666 | |
D28875.1 | - | 10 | 960 | |||
FQ212499.1 | - | 10 | 978 | |||
FQ214222.1 | - | 10 | 975 | |||
FQ214492.1 | - | 10 | 977 | |||
FQ214649.1 | - | 10 | 978 | |||
FQ214673.1 | - | 10 | 977 | |||
FQ215195.1 | - | 10 | 976 | |||
FQ215581.1 | - | 10 | 978 | |||
FQ215622.1 | - | 10 | 979 | |||
FQ217546.1 | - | 10 | 978 | |||
FQ219932.1 | - | 11 | 959 | |||
FQ219983.1 | - | 10 | 977 | |||
FQ220003.1 | - | 10 | 977 | |||
FQ220043.1 | - | 10 | 975 | |||
FQ220046.1 | - | 10 | 977 | |||
FQ220186.1 | - | 10 | 975 | |||
FQ220279.1 | - | 8 | 801 | |||
FQ220287.1 | - | 10 | 979 | |||
FQ220507.1 | - | 10 | 985 | |||
FQ220547.1 | - | 10 | 977 | |||
FQ220827.1 | - | 10 | 977 | |||
FQ220880.1 | - | 10 | 977 | |||
FQ220901.1 | - | 10 | 977 | |||
FQ224996.1 | - | 9 | 848 | |||
FQ225033.1 | - | 10 | 979 | |||
FQ225092.1 | - | 10 | 973 | |||
FQ225220.1 | - | 10 | 956 | |||
FQ225486.1 | - | 10 | 959 | |||
FQ225609.1 | - | 10 | 977 | |||
FQ225618.1 | - | 10 | 975 | |||
FQ225644.1 | - | 6 | 644 | |||
FQ225688.1 | - | 10 | 977 | |||
FQ225694.1 | - | 10 | 979 | |||
FQ225722.1 | - | 10 | 976 | |||
FQ225884.1 | - | 10 | 975 | |||
FQ225913.1 | - | 10 | 977 | |||
FQ225914.1 | - | 10 | 979 | |||
FQ225965.1 | - | 10 | 977 | |||
FQ226299.1 | - | 10 | 959 | |||
FQ226354.1 | - | 10 | 977 | |||
FQ226360.1 | - | 10 | 977 | |||
FQ226599.1 | - | 10 | 978 | |||
FQ226731.1 | - | 10 | 977 | |||
FQ226746.1 | - | 10 | 975 | |||
FQ227727.1 | - | 10 | 957 | |||
FQ227749.1 | - | 10 | 977 | |||
FQ228032.1 | - | 10 | 957 | |||
FQ229230.1 | - | 10 | 977 | |||
FQ229283.1 | - | 10 | 959 | |||
FQ229299.1 | - | 10 | 960 | |||
FQ229316.1 | - | 10 | 977 | |||
FQ230029.1 | - | 10 | 977 | |||
FQ230595.1 | - | 10 | 959 | |||
FQ230667.1 | - | 10 | 978 | |||
FQ230705.1 | - | 10 | 975 | |||
FQ230778.1 | - | 10 | 977 | |||
FQ230864.1 | - | 10 | 977 | |||
FQ230993.1 | - | 10 | 977 | |||
FQ231067.1 | - | 10 | 977 | |||
FQ231444.1 | - | 10 | 976 | |||
FQ231543.1 | - | 10 | 977 | |||
FQ231632.1 | - | 10 | 977 | |||
FQ231815.1 | - | 10 | 959 | |||
FQ231918.1 | - | 10 | 959 | |||
FQ231975.1 | - | 10 | 959 | |||
FQ232042.1 | - | 10 | 976 | |||
FQ232191.1 | - | 10 | 977 | |||
FQ232271.1 | - | 10 | 974 | |||
FQ232364.1 | - | 10 | 979 | |||
FQ232445.1 | - | 10 | 975 | |||
FQ232464.1 | - | 10 | 977 | |||
FQ232710.1 | - | 10 | 975 | |||
FQ232838.1 | - | 10 | 977 | |||
FQ232847.1 | - | 10 | 977 | |||
FQ232862.1 | - | 10 | 975 | |||
FQ232869.1 | - | 10 | 977 | |||
FQ232965.1 | - | 10 | 977 | |||
FQ232977.1 | - | 10 | 977 | |||
FQ232986.1 | - | 10 | 977 | |||
FQ233178.1 | - | 10 | 977 | |||
FQ233283.1 | - | 10 | 956 | |||
FQ233669.1 | - | 10 | 977 | |||
FQ233698.1 | - | 10 | 959 | |||
FQ233777.1 | - | 10 | 975 | |||
FQ233810.1 | - | 10 | 979 | |||
FQ233845.1 | - | 10 | 977 | |||
FQ233850.1 | - | 10 | 978 | |||
FQ234037.1 | - | 10 | 977 | |||
FQ234108.1 | - | 10 | 977 | |||
FQ234199.1 | - | 10 | 977 | |||
FQ234418.1 | - | 10 | 977 | |||
FQ234523.1 | - | 10 | 964 | |||
FQ234589.1 | - | 10 | 975 | |||
FQ234657.1 | - | 10 | 959 | |||
FQ234685.1 | - | 10 | 977 | |||
FQ234766.1 | - | 10 | 975 | |||
FQ235008.1 | - | 10 | 979 | |||
FQ235049.1 | - | 10 | 959 | |||
Y13714.1 | - | 11 | 943 |