Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013110.1 | NP_001013128.1 | 11, 12 | 1352 | ||
GenBank mRNA | AF476964.1 | - | 11, 12 | 1383 | ||
AY327504.1 | - | 11, 12 | 1321 | |||
BC087021.1 | - | 11, 12 | 1352 | 7132382 | ||
D38380.1 | - | 11, 12 | 1351 | |||
FQ209414.1 | - | 4, 5 | 316 | |||
FQ209524.1 | - | 4, 5 | 411 | |||
FQ209552.1 | - | 2, 3 | 131 | |||
FQ209673.1 | - | 5, 6 | 519 | |||
FQ209674.1 | - | 5, 6 | 576 | |||
FQ209794.1 | - | 3, 4 | 204 | |||
FQ211385.1 | - | 2, 3 | 128 | |||
FQ213988.1 | - | 1, 2 | 37 | |||
FQ214362.1 | - | 4, 5 | 429 | |||
FQ218210.1 | - | 3, 4 | 182 | |||
FQ219264.1 | - | 5, 6 | 455 | |||
FQ219349.1 | - | 3, 4 | 185 | |||
FQ219724.1 | - | 5, 6 | 483 | |||
X77158.1 | - | 11, 12 | 1329 |
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY325214.1 | - | 1319 | |||
AY325230.1 | - | 1319 | ||||
AY325241.1 | - | 1319 |