Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052801.1 | NP_434688.1 | 2, 3 | 472 | ||
GenBank mRNA | S80345.1 | - | 2, 3 | 413 | ||
U14746.1 | - | 2, 3 | 472 |