Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031534.2 | NP_113722.2 | 8 | 1085 | ||
XM_006234620.1 | XP_006234682.1 | 7 | 1427 | |||
XM_006234621.2 | XP_006234683.1 | 8 | 1556 | |||
XM_006234622.2 | XP_006234684.1 | 7 | 1505 | |||
GenBank mRNA | X69716.1 | - | 8 | 1085 |