Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052803.2 | NP_434690.1 | 17 | 3567 | ||
XM_006256995.2 | XP_006257057.1 | 18 | 3734 | |||
XM_006256997.2 | XP_006257059.1 | 18 | 3731 | |||
XM_008773334.1 | XP_008771556.1 | 18 | 3675 | |||
GenBank mRNA | U59245.1 | - | 17 | 3572 |