Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030990.2 | NP_112252.1 | 6, 7 | 903 | ||
XM_006257293.2 | XP_006257355.1 | 9, 10 | 1185 | |||
XM_008773401.1 | XP_008771623.1 | 9, 10 | 1289 | |||
GenBank mRNA | BC093596.1 | - | 5, 6 | 764 | 7314758 | |
FQ211530.1 | - | 6, 7 | 876 | |||
FQ212519.1 | - | 5, 6 | 512 | |||
FQ212680.1 | - | 5, 6 | 617 | |||
FQ213540.1 | - | 5, 6 | 617 | |||
FQ213548.1 | - | 5, 6 | 618 | |||
FQ213841.1 | - | 5, 6 | 617 | |||
FQ213943.1 | - | 5, 6 | 617 | |||
FQ214012.1 | - | 5, 6 | 617 | |||
FQ214050.1 | - | 5, 6 | 617 | |||
FQ214153.1 | - | 5, 6 | 618 | |||
M11185.1 | - | 6, 7 | 878 | |||
M14126.1 | - | 3, 4 | 231 | |||
M25888.1 | - | 4, 5 | 443 | |||
X02809.1 | - | 6, 7 | 807 |