Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031572.1 | NP_113760.1 | 5 | 688 | ||
GenBank mRNA | FQ209627.1 | - | 2 | 127 | ||
FQ219038.1 | - | 3 | 369 | |||
FQ219491.1 | - | 4 | 563 | |||
FQ219720.1 | - | 2 | 158 | |||
J03786.1 | - | 5 | 688 |